Learn More
SGK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28454-1-AP-150UL
 View more versions of this product
Product Code. 16876145
  • 4652.90 SEK / 150µL
Description
The SGK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SGK1. This antibody recognizes human antigen. The SGK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SGK1
Polyclonal
Unconjugated
150 μL
6446
IgG
Western Blot, Immunohistochemistry
450 μg/mL
Rabbit
Primary
Human
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SGK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL