missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
SGTA Mouse anti-Human, Clone: 1.90E+09, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59426-100UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SGTA Monoclonal specifically detects SGTA in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifikationer
| SGTA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1036, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6449 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1900000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| SGTA | |
| Recombinant human SGTA (1-313aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering