Learn More
SHARPIN Mouse anti-Human, Unconjugated, Clone: 3A9E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67177-1-IG-150UL
 View more versions of this product
Product Code. 16879665
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The SHARPIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SHARPIN. This antibody recognizes human antigen. The SHARPIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SHARPIN
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3A9E2
Unconjugated
150 μL
81858
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SHARPIN Mouse anti-Human, Unconjugated, Clone: 3A9E2, Proteintech > 150 μL