Learn More
SHARPIN Mouse anti-Human, Unconjugated, Clone: 3A9E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67177-1-IG-20UL
 View more versions of this product
Product Code. 16889665
  • 1645.54 SEK / 20µL
Description
The SHARPIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SHARPIN. This antibody recognizes human antigen. The SHARPIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SHARPIN
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3A9E2
Unconjugated
20 μL
81858
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SHARPIN Mouse anti-Human, Unconjugated, Clone: 3A9E2, Proteintech > 20 μL