missing translation for 'onlineSavingsMsg'
Learn More

SHCBP1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27108-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885395

  • 1656.73 SEK / 20µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a Src homology 2 domain-binding protein 1-like protein. The encoded protein interacts with heat shock 70 kDa protein 2 and may be involved in maintaining spindle integrity during meiosis. This gene is located in region of chromoso0me 1 encompassing a prostate cancer susceptibility locus.
Specifications

Specifications

SHCBP1L
Polyclonal
Unconjugated
SHCBP1L
C1orf14, GE36, SHCBP1L
Rabbit
Antigen Affinity Chromatography
RUO
289101, 71836, 81626
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TTP0, Q9BZQ2
Shcbp1l
SHCBP1L Fusion Protein Ag25903
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers