Learn More
Trajan Scientific and Medical™ Shimadzu GC-2010 Liners
Provides excellent analysis for a wide range of compounds.
3055.00 SEK - 4450.00 SEK
Specifications
Description | GC-2010 Liners |
---|---|
Diameter (Metric) Inner | 1.5 mm |
Diameter (Metric) Outer | 3.1 mm |
Liner Material | Borosilicate Glass |
Product Code | Brand | Product Type Specs | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type Specs | Price | Quantity & Availability | |||||
15219013
|
Trajan Scientific and Medical™
0920425 |
Inlet liners PTV 3 baffle |
4450.00 SEK
Pack of 5 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
15239013
|
Trajan Scientific and Medical™
092292 |
Straight LVI Liner with Wool |
3055.00 SEK
Pack of 5 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Shimadzu instruments
Specifications
GC-2010 Liners | |
1.5 mm | |
3.1 mm | |
Borosilicate Glass |