Learn More
Trajan™ SilTite™ FingerTite Ferrule System Adapters
For gas chromatography systems
Brand:  Trajan™ 0736102
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15208953
  • 1025.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Adapter
Agilent MS Adaptor
Agilent GCs