Learn More
Silver Nitrate Solution 1000g/L, for COD, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Color | Colorless |
MDL Number | 3414 |
Packaging | Plastic Coated Glass bottle |
Quantity | 500 mL |
Physical Form | Liquid |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Water analysis
File size: 7.3 MB
Product Content Correction
Product Title
Silver Nitrate Solution 1000g/L, for COD, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction