Learn More
Silver Nitrate Solution 1M (1N), Standard Solution Ready To Use for Volumetric Analysis, Fisher Chemical™ Product Code.: 10725911
Silver Nitrate Solution 1M (1N), Standard Solution Ready To Use for Volumetric Analysis, Fisher Chemical™
Product Code. p-7073174 Shop All Fisher Chemical Products
1 L, 1 L
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
Product Code. 10725911
Brand: Fisher Chemical J/7320/PB15
Request Bulk or Custom Format
Chemical Identifiers
CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O
Specifications
Color Colorless
IUPAC Name silver(1+) nitrate
Packaging HDPE plastic bottle
pH 5.4 to 6.4
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Silver nitrate, 1M (1N) aqueous solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Product Content Correction
Product Title
Silver Nitrate Solution 1M (1N), Standard Solution Ready To Use for Volumetric Analysis, Fisher Chemical™ >