Learn More
Socorex™ Acura™ Single-channel Macropipette Volumetric Module
Increase the versatility of your Acura™ electronic macropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Macropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel macropipetting needs.
1240.00 SEK
Specifications
For Use With | Acura™ Electro™ 926 MicroPipette |
---|---|
Number of Channels | 1 |
Product Line | Acura |
Type | Electronic Pipette Volumetric Module |
For Use With (Equipment) | Acura™ Electro 926 Micropipette |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10265282
|
Socorex™
800.5000 |
0.25 to 5 mL |
1240.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
10632203
|
Socorex™
800.10000 |
0.5 to 10 mL |
1240.00 SEK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Acura™ Electro™ 926 MicroPipette | |
Acura | |
Acura™ Electro 926 Micropipette |
1 | |
Electronic Pipette Volumetric Module | |
1 Each |