missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAMF6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22711-1-AP-150UL
This item is not returnable.
View return policy
Description
Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.Specifications
SLAMF6 | |
Polyclonal | |
Unconjugated | |
SLAMF6 | |
Activating NK receptor, CD352, KALI, KALIb, Ly108, NK T B antigen, NTB A, NTBA, SF2000, SLAM family member 6, SLAMF6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114836 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96DU3 | |
SLAMF6 | |
SLAMF6 Fusion Protein Ag18517 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLAMF6 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction