missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAMF7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12905-1-AP-150UL
This item is not returnable.
View return policy
Description
SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.Specifications
SLAMF7 | |
Polyclonal | |
Unconjugated | |
SLAMF7 | |
19A, CD2 subset 1, CD319, CRACC, CS1, Membrane protein FOAP 12, Novel Ly9, Protein 19A, SLAM family member 7, SLAMF7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57823 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NQ25 | |
SLAMF7 | |
SLAMF7 Fusion Protein Ag3804 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLAMF7 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction