missing translation for 'onlineSavingsMsg'
Learn More

SLC16A12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20553-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890105

  • 1355.00 SEK / 20µL
Estimated Shipment: 28-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to SLC16A12.

This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
Specifications

Specifications

SLC16A12
Polyclonal
Unconjugated
SLC16A12
CJMG, MCT 12, MCT12, Monocarboxylate transporter 12, SLC16A12
Rabbit
Antigen Affinity Chromatography
RUO
240638, 309525, 387700
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZSM3, Q8BGC3
SLC16A12
Peptide
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers