missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24901-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
SLC22A23 | |
Polyclonal | |
Unconjugated | |
SLC22A23 | |
BOCT2, C6orf85, SLC22A23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63027, 64559, 73102 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1A5C7, Q3UHH2, Q9QZG1 | |
SLC22A23 | |
SLC22A23 Fusion Protein Ag19036 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |