Learn More
SLC22A7 Mouse anti-Human, Unconjugated, Clone: 3D3C3, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SLC22A7 |
---|---|
Clone | 3D3C3 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880126
|
Proteintech
67479-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870126
|
Proteintech
67479-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SLC22A7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC22A7. This antibody recognizes human antigen. The SLC22A7 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SLC22A7 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
10864 | |
Primary |
3D3C3 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.