Learn More
SLC22A9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27998-1-AP-20UL
Description
The SLC22A9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC22A9. This antibody recognizes human, mouse antigen. The SLC22A9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SLC22A9 | |
Polyclonal | |
Unconjugated | |
20 μL | |
114571 | |
IgG |
Western Blot, Immunohistochemistry | |
500 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |