Learn More
SLC25A12 Mouse anti-Human, Unconjugated, Clone: 1F2B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67467-1-IG-20UL
Description
The SLC25A12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC25A12. This antibody recognizes human, mouse antigen. The SLC25A12 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SLC25A12 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
1F2B10 | |
Unconjugated | |
20 ÎĽL | |
8604 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.