Learn More
SLC25A12 Mouse anti-Human, Unconjugated, Clone: 1F2B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67467-1-IG-20UL
 View more versions of this product
Product Code. 16880106
  • 1645.54 SEK / 20µL
Description
The SLC25A12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC25A12. This antibody recognizes human, mouse antigen. The SLC25A12 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SLC25A12
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry, ELISA
1F2B10
Unconjugated
20 ÎĽL
8604
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SLC25A12 Mouse anti-Human, Unconjugated, Clone: 1F2B10, Proteintech > 20 ÎĽL