missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SLC25A3 Recombinant Protein

Recombinant protein for SLC25A3 (human) gene

Brand:  Abnova™ H00005250-P01.25ug

 View more versions of this product

Product Code. 16189021

  • 6845.00 SEK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The encoded protein catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. It contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family.

  • Human SLC25A3 full-length ORF ( AAH00998, 1 a.a. - 361 a.a.) recombinant protein with GST-tag at N-terminal
  • Description: solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3
  • Theoretical molecular weight: 65.45kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer

Sequence: MFSSVAHLARANPFNTPHLQLVHDGLGDLRSSSPGPTGQPRRPRNLAAAAVEEYSCEFGSAKYYALCGFGGVLSCGLTHTAVVPLDLVKCRMQVDPQKYKGIFNGFSVTLKEDGVRGLAKGWAPTFLGYSMQGLCKFGFYEVFKVLYSNMLGEENTYLWRTSLYLAASASAEFFADIALAPME AAKVRIQTQPGYANTLRDAAPKMYKEEGLKAFYKGVAPLWMRQIPYTMMKFACCERTVEALYKFVVPKPRSECSKPEQLVVTFVAGYIAGVFCAIVSHPADSVVSVLNKEKGSSASLVLKRLGFKGVWKGLFARIIMIGTLTALQWFIYDSVKVYFRLPRPPPPEMPESLKKKLGLTQ

Best use within three months from the date of receipt.

ELISA, western blotting (recombinant protein), antibody production, protein array

Specifications

Specifications

Antibody Production, ELISA, Protein Array, Western Blot
65.4
8
Glutathione Sepharose 4 Fast Flow
25 μg
-80°C
Recombinant
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human SLC25A3 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers