missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SLC25A34 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897464
|
Proteintech
17557-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887464
|
Proteintech
17557-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins.Specifications
SLC25A34 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A2ADF7, Q5XIF9, Q6PIV7 | |
284723, 298606, 384071 | |
SLC25A34 Fusion Protein Ag11719 | |
Primary | |
-20°C | |
SLC25A34 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RP11 169K16.2, SLC25A34 | |
SLC25A34 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC25A34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech