missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC37A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20612-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.Specifications
SLC37A4 | |
Polyclonal | |
Unconjugated | |
SLC37A4 | |
G6PT, G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, SLC37A4, TRG 19, TRG19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14385, 2542, 29573 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43826 | |
SLC37A4 | |
SLC37A4 Fusion Protein Ag14660 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC37A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction