Learn More
SLC38A4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20857-1-AP-20UL
 View more versions of this product
Product Code. 16890435
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
SLC38A4 is found predomitly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
SLC38A4
Polyclonal
Unconjugated
SLC38A4
Amino acid transporter A3, ATA3, NAT3, PAAT, SLC38A4, SNAT4
Rabbit
Antigen Affinity Chromatography
RUO
55089
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969I6
SLC38A4
SLC38A4 Fusion Protein Ag14827
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC38A4 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated