missing translation for 'onlineSavingsMsg'
Learn More

SLC45A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen SLC45A2
Concentration 0.15 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16826853
View Documents
Proteintech
10453-1-AP-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16816853
View Documents
Proteintech
10453-1-AP-150UL
150 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

SLC45A2 is a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, ′B, ′ encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. The protein encoded by this gene encodes a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, ′B, ′ encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

SLC45A2
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
P58355, Q9UMX9
22293, 310152, 51151
SLC45A2 Fusion Protein Ag0699
Primary
-20°C
SLC45A2
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
1A1, AIM1, MATP, Melanoma antigen AIM1, Protein AIM 1, SHEP5, SLC45A2
SLC45A2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SLC45A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.