Learn More
SLC7A6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13823-1-AP-150UL
 View more versions of this product
Product Code. 16882214
  • 4652.90 SEK / 150µL
Description
The SLC7A6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC7A6. This antibody recognizes human, mouse antigen. The SLC7A6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SLC7A6
Polyclonal
Unconjugated
150 μL
9057
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL
Rabbit
Primary
Human, Mouse
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SLC7A6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL