missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SLC9A9 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882074
|
Proteintech
13718-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872074
|
Proteintech
13718-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.Specifications
SLC9A9 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8BZ00, Q8IVB4 | |
285195, 331004, 363115 | |
SLC9A9 Fusion Protein Ag4660 | |
Primary | |
-20°C | |
SLC9A9 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Na(+)/H(+) exchanger 9, Nbla00118, NHE 9, NHE9, SLC9A9, Sodium/hydrogen exchanger 9 | |
Slc9a9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC9A9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech