Learn More
SLU7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12050-1-AP-20UL
 View more versions of this product
Product Code. 16879533
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
Specifications
Specifications
SLU7
Polyclonal
Unconjugated
SLU7
9G8, hSlu7, Pre mRNA splicing factor SLU7, SLU7
Rabbit
Antigen Affinity Chromatography
RUO
10569, 193116, 303057
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95391, Q80ZG5, Q8BHJ9
SLU7
SLU7 Fusion Protein Ag2680
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLU7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated