Learn More
SLU7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12050-1-AP-20UL
 View more versions of this product
Product Code. 16879533
  • 1645.54 SEK / 20µL
Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
Specifications
Specifications
SLU7
Polyclonal
Unconjugated
SLU7
9G8, hSlu7, Pre mRNA splicing factor SLU7, SLU7
Rabbit
Antigen Affinity Chromatography
RUO
10569, 193116, 303057
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95391, Q80ZG5, Q8BHJ9
SLU7
SLU7 Fusion Protein Ag2680
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SLU7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated