Läs mer
Smad7 Antibody - Azide Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-56688SS
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Smad7 Polyclonal specifically detects Smad7 in Human, Mouse, Sheep samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifikationer
| Smad7 | |
| Polyclonal | |
| Unconjugated | |
| Sterile - filtered PBS with No Preservative | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 4092 | |
| Human, Mouse, Sheep | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 4 - 6 ug/ml | |
| NP_005895 | |
| SMAD7 | |
| This antibody was raised against two peptides made from within amino acids 10 and 50 of human SMAD7. This sequence is 100% or highly conserved between human and multiple species. | |
| 0.025 mg | |
| Primary | |
| In A549, an approx. 45 kDa band is observed. This may cross-react with SMAD6 depending on cell or tissue-specific expression. Cross reacts with Human, Mouse and Rat. Predicted to react with Chimpanzee, Dog, Pufferfish, Zebrafish and Xenopus. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering