missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Smad7 Antibody - Azide Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
2165.00 SEK
Specifikationer
| Antigen | Smad7 |
|---|---|
| Koncentration | 1 mg/ml |
| Utspädning | Western Blot 4 - 6 ug/ml |
| Användningsområden | Western Blot, Immunohistochemistry (Paraffin) |
| Klassificering | Polyclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
18047163
|
Novus Biologicals
NB100-56688SS |
0.025 mg |
2165.00 SEK
25µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Smad7 Polyclonal specifically detects Smad7 in Human, Mouse, Sheep samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifikationer
| Smad7 | |
| Western Blot 4 - 6 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Sterile - filtered PBS with No Preservative | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
| SMAD7 | |
| IgG | |
| Protein G purified | |
| In A549, an approx. 45 kDa band is observed. This may cross-react with SMAD6 depending on cell or tissue-specific expression. Cross reacts with Human, Mouse and Rat. Predicted to react with Chimpanzee, Dog, Pufferfish, Zebrafish and Xenopus. |
| 1 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Sheep | |
| NP_005895 | |
| 4092 | |
| This antibody was raised against two peptides made from within amino acids 10 and 50 of human SMAD7. This sequence is 100% or highly conserved between human and multiple species. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel