missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26613-1-AP-20UL
This item is not returnable.
View return policy
Description
SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.Specifications
SMARCA2 | |
Polyclonal | |
Unconjugated | |
SMARCA2 | |
ATP dependent helicase SMARCA2, BAF190, BAF190B, BRG1 associated factor 190B, BRM, hBRM, hSNF2a, Protein brahma homolog, SMARCA2, SMARCA2-Specific, SNF2, SNF2 alpha, SNF2A, SNF2L2, SNF2LA, Sth1p, SWI2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6595, 67155 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51531, Q6DIC0 | |
SMARCA2 | |
SMARCA2-Specific Fusion Protein Ag25157 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMARCA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction