Learn More
SMCHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25589-1-AP-20UL
 View more versions of this product
Product Code. 16884145
  • 1645.54 SEK / 20µL
Description
This gene encodes a protein which contains a hinge region domain found in members of the SMC (structural maintece of chromosomes) protein family.
Specifications
Specifications
SMCHD1
Polyclonal
Unconjugated
SMCHD1
KIAA0650, SMCHD1
Rabbit
Antigen Affinity Chromatography
RUO
23347, 74355
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NHR9, Q6P5D8
Smchd1
SMCHD1 Fusion Protein Ag22293
20 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SMCHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated