Learn More
SMCHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25589-1-AP-20UL
 View more versions of this product
Product Code. 16884145
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a protein which contains a hinge region domain found in members of the SMC (structural maintece of chromosomes) protein family.
Specifications
Specifications
SMCHD1
Polyclonal
Unconjugated
SMCHD1
KIAA0650, SMCHD1
Rabbit
Antigen Affinity Chromatography
RUO
23347, 74355
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NHR9, Q6P5D8
Smchd1
SMCHD1 Fusion Protein Ag22293
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMCHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated