Learn More
SMN-Exon7 Mouse anti-Human, CoraLite 594, Clone: 3A8G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60255-100UL
Description
The SMN-Exon7 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human SMN-Exon7. This antibody recognizes human, mouse antigen. The SMN-Exon7 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
SMN-Exon7 | |
Monoclonal | |
CoraLite 594 | |
100 ÎĽL | |
6606 | |
IgG1 |
Immunofluorescence | |
3A8G11 | |
Mouse | |
Primary | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.