missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMTNL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SMTNL2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883594
|
Proteintech
14782-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873594
|
Proteintech
14782-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SMTNL2 | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q2TAL5, Q8CI12 | |
276829, 342527, 679629 | |
SMTNL2 Fusion Protein Ag6395 | |
Primary | |
-20°C | |
SMTNL2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
smoothelin like 2, Smoothelin like protein 2, SMTNL2 | |
Smtnl2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMTNL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech