Learn More
SMYD4 Mouse anti-Human, Unconjugated, Clone: 2F4B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66874-1-IG-150UL
 View more versions of this product
Product Code. 16879135
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The SMYD4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMYD4. This antibody recognizes human antigen. The SMYD4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SMYD4
Monoclonal
920 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2F4B7
Unconjugated
150 μL
114826
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMYD4 Mouse anti-Human, Unconjugated, Clone: 2F4B7, Proteintech > 150 μL