Learn More
SNAI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12129-1-AP-150UL
 View more versions of this product
Product Code. 16889653
  • 4652.90 SEK / 150µL
Description
The SNAI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNAI2. This antibody recognizes human, mouse antigen. The SNAI2 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
Specifications
SNAI2
Polyclonal
Unconjugated
150 μL
6591
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
333 μg/mL
Rabbit
Primary
Human, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SNAI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL