missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD2 Mouse anti-Human, Clone: 1F12B2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SNRPD2 |
---|---|
Clone | 1F12B2 |
Concentration | 1.27 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897685
|
Proteintech
66111-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887685
|
Proteintech
66111-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SNRPD2 | |
1.27 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Sm D2, SMD2, snRNP core protein D2, SNRPD2 | |
SNRPD2 | |
IgG2b | |
Protein A | |
Antibody |
1F12B2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P62316 | |
6633 | |
SNRPD2 Fusion Protein Ag7115 | |
Primary | |
-20°C | |
SNRPD2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SNRPD2 Mouse anti-Human, Clone: 1F12B2, Proteintech