Learn More
SNRPN Mouse anti-Human, Unconjugated, Clone: 1H2F4, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SNRPN |
---|---|
Clone | 1H2F4 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889815
|
Proteintech
67280-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879815
|
Proteintech
67280-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SNRPN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNRPN. This antibody recognizes human, mouse, rat antigen. The SNRPN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SNRPN | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
6638 | |
Primary |
1H2F4 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.