missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNUPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15358-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.Specifications
SNUPN | |
Polyclonal | |
Unconjugated | |
SNUPN | |
KPNBL, RNA U transporter 1, RNUT1, SNUPN, snurportin 1, Snurportin1, SPN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10073, 316108, 66069 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95149, Q68FP5, Q80W37 | |
Snupn | |
SNUPN Fusion Protein Ag7604 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SNUPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction