Learn More
Sodium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
1360.00 SEK - 3205.00 SEK
Chemical Identifiers
CAS | 7440-23-5, 7732-18-5, 7697-37-2 |
---|---|
Molecular Formula | Na |
Molecular Weight (g/mol) | 22.99 |
MDL Number | MFCD00085307 |
InChI Key | KEAYESYHFKHZAL-UHFFFAOYSA-N |
PubChem CID | 5360545 |
ChEBI | CHEBI:26708 |
IUPAC Name | sodium |
SMILES | [Na] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10375692
|
SPEX Certiprep
PLNA2-3Y |
125 mL | Triple-leached LDPE bottle |
1360.00 SEK
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10652333
|
SPEX Certiprep
PLNA2-3X |
500 mL | Triple-leached LDPE bottle |
3205.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Na | 10,000ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-23-5, 7732-18-5, 7697-37-2 | |
22.99 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
CHEBI:26708 | |
[Na] |
Na | |
MFCD00085307 | |
5360545 | |
sodium |
Specifications
7440-23-5, 7732-18-5, 7697-37-2 | |
Na | |
AA, ICP | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
[Na] | |
5% HNO3 | |
5360545 | |
Assurance |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
MFCD00085307 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Certified Reference Material | |
sodium | |
22.99 | |
CHEBI:26708 | |
Sodium Standard |
Safety and Handling
Harmful in contact with skin. Causes serious eye irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
- 10,000
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : nitric acid, Sodiumfrom Sodium carbonate
Product intended for laboratory use only