Learn More
Memmert™ Software Conforming to FDA AtmoCONTROL
Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Brand:  Memmert™ ICO150FDAQ1
 View more versions of this product
Product Code. 17169964
  • 30290.00 SEK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
  • Base licence for reading the data logging of one unit (only for units with TwinDISPLAY)
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Only for Units with TwinDISPLAY
ICO150
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Software Conforming to FDA AtmoCONTROL > ICO150