missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol dehydrogenase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15881-1-AP-20UL
This item is not returnable.
View return policy
Description
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.Specifications
Sorbitol dehydrogenase | |
Polyclonal | |
Unconjugated | |
SORD | |
L iditol 2 dehydrogenase, SDH, Sorbitol dehydrogenase, SORD, SORD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20322, 24788, 6652 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27867, Q00796, Q64442 | |
SORD | |
Sorbitol dehydrogenase Fusion Protein Ag8651 | |
20 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sorbitol dehydrogenase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction