missing translation for 'onlineSavingsMsg'
Learn More

SOX7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23925-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882525

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may play a role in tumorigenesis. A similar protein in mice is involved in the regulation of the wingless-type MMTV integration site family (Wnt) pathway.
Specifications

Specifications

SOX7
Polyclonal
Unconjugated
SOX7
SOX7, Transcription factor SOX 7
Rabbit
Antigen Affinity Chromatography
RUO
20680, 83595
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P40646, Q9BT81
SOX7
SOX7 Fusion Protein Ag10323
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers