Learn More
Abnova™ SP7/CEN12p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0215.100uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MGC126598,OSX,osterix | |
121340 | |
100 μL |
Tube | |
Sp7 transcription factor | |
SP7 | |
DNA/RNA | |
Liquid |