missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SP7/CEN12p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
19880.00 SEK - 32740.00 SEK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | Sp7 transcription factor |
Gene Alias | MGC126598,OSX,osterix |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073345
|
Abnova™
FG0215.100UL |
100 μL |
19880.00 SEK
100µL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083345
|
Abnova™
FG0215.200UL |
200 μL |
32740.00 SEK
200µL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MGC126598,OSX,osterix | |
121340 | |
Liquid |
Tube | |
Sp7 transcription factor | |
SP7 | |
DNA/RNA |