missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16294-1-AP-150UL
This item is not returnable.
View return policy
Description
SPAG7 gene ontology annotations related to this gene include nucleic acid binding.Specifications
SPAG7 | |
Polyclonal | |
Unconjugated | |
SPAG7 | |
ACRP, FSA 1, SPAG7, sperm associated antigen 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216873, 303260, 9552 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75391, Q7TNE3 | |
SPAG7 | |
SPAG7 Fusion Protein Ag9471 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SPAG7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction