missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25861-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
SPATC1 | |
Polyclonal | |
Unconjugated | |
SPATC1 | |
SPATA15, SPATC1, SPERIOLIN, SPRN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
375686, 74281 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q148B6, Q76KD6 | |
SPATC1 | |
SPATC1 Fusion Protein Ag12157 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |