missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SPATC1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824425
|
Proteintech
25861-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814425
|
Proteintech
25861-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SPATC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q148B6, Q76KD6 | |
375686, 74281 | |
SPATC1 Fusion Protein Ag12157 | |
Primary | |
-20°C | |
SPATC1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SPATA15, SPATC1, SPERIOLIN, SPRN | |
SPATC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |