missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDEF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11467-1-AP-150UL
This item is not returnable.
View return policy
Description
PDEF (prostate-derived Ets factor) is a 37.5- kDa protein that acts as an androgen-independent transcriptional activator for the PSA (prostate specific antigen) promoter. It also interacts directly with the DNA binding domain of androgen receptor and enhances androgen-mediated activation of the PSA promoter. PDEF was shown to have transcript levels 192-fold higher in the peripheral blood of some breast cancer patients in comparison with normal individuals. PDEF mRNA is also frequently over-expressed in human breast tumors as well as in human ovarian tumors. In ovarian tumors, PDEF expression seems to down-modulate maligt potential, and thus provides a rationale to screen for drugs that induce PDEF expression in epithelial ovarian tumors.Specifications
SPDEF | |
Polyclonal | |
Unconjugated | |
SPDEF | |
PDEF, Prostate derived Ets factor, Prostate specific Ets, PSE, RP11 375E1__A.3, SPDEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25803, 30051, 689210 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95238, Q9WTP3 | |
SPDEF | |
SPDEF Fusion Protein Ag2020 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SPDEF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction