missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDYA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SPDYA |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837834
|
Proteintech
17836-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16827834
|
Proteintech
17836-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Speedy A, also known as SPDYA, SPDY1, Ringo3 or SPY1, is a 313 amino acid protein that localizes to the nucleus and belongs to the speedy/ringo family. Expressed at high levels in testis and at lower levels in brain, kidney, heart, bone marrow, colon, lung, liver spleen and placenta, Speedy A functions to regulate the G1/S phase transition of the cell cycle, specifically by binding to and activating Cdc2 p34, Cdk2 and p27. Additionally, Speedy A mediates cell survival during DNA damage, suggesting that Speedy A plays a role in proper cell cycle progression throughout the life of the cell. Multiple isoforms of Speedy A exist due to alternative splicing events. The gene encoding Speedy A maps to human chromosome 2, which encodes over 1,400 genes and comprises nearly 8% of the human genome.Specifications
SPDYA | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q5IBH7, Q5MJ70, Q8R496 | |
192209, 245711, 70891 | |
SPDYA Fusion Protein Ag12041 | |
Primary | |
-20°C | |
SPDYA |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hSpy/Ringo A, RINGO A, Ringo3, SPDY1, SPDYA, Speedy 1, Speedy protein A, SPY1 | |
Spdya | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SPDYA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech